Learn More
Medchemexpress LLC HY-114758 10mg Medchemexpress, Pyridoxal isonicotinoyl hydrazone CAS:737-86-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11475810MG
Encompass_Preferred
Medchemexpress, HY-114758 10mg Pyridoxal isonicotinoyl hydrazone CAS:737-86-0 Pyridoxal isonicotinoyl hydrazone (PIH) is a lipophilic, tridentate Fe-chelating agent that shows high Fe chelation efficacy [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2499