Learn More
Medchemexpress LLC HY-B1744 100mg Medchemexpress, Pyridoxal phosphate CAS:54-47-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1744100MG
Encompass_Preferred
Medchemexpress, HY-B1744 100mg Pyridoxal phosphate CAS:54-47-7 Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases , and is used for the treatment of tardive dyskinesia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2998