Learn More
Medchemexpress LLC Pyridoxal phosphate | 54-47-7 | 99.67% | 50 MG
SDP
Catalog No. 5000411932
Encompass_Preferred
Pyridoxal phosphate is the active form of vitamin B6. It acts as an inhibitor of reverse transcriptases and is used for the treatment of tardive dyskinesia.
  • Target: Reverse transcriptase, endogenous metabolite, ERK, amyloid-β.
  • Research areas: Neurological disease, metabolic disease, cancer.
  • Category: Human gut microbiota metabolites, immune system disorder, microorganisms, ketones, aldehydes, acids, disease markers, endocrine diseases, nervous system disorder, endogenous metabolite.
  • Available as an analytical standard for research and analytical applications.
  • Available as a deuterium-labeled compound.
  • Verification: 3 publications.
Catalog No. 50-004-11932 Supplier Medchemexpress LLC Supplier No. HYB1744R50MG
Pyridoxal phosphate is the active form of vitamin B6. It acts as an inhibitor of reverse transcriptases and is used for the treatment of tardive dyskinesia.
  • Target: Reverse transcriptase, endogenous metabolite, ERK, amyloid-β.
  • Research areas: Neurological disease, metabolic disease, cancer.
  • Category: Human gut microbiota metabolites, immune system disorder, microorganisms, ketones, aldehydes, acids, disease markers, endocrine diseases, nervous system disorder, endogenous metabolite.
  • Available as an analytical standard for research and analytical applications.
  • Available as a deuterium-labeled compound.
  • Verification: 3 publications.