Learn More
Medchemexpress LLC Pyridoxal phosphate | 54-47-7 | 99.95% | 500 UG
SDP
Catalog No. 5000413477
Encompass_Preferred
Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia. This product is for research use only.
  • Severely inhibits DNA Polymerase and RNase H, leading to an immediate reduction in DNA synthesis.
  • Conjugation to pyridoxal phosphate fully inhibits NEIL2.
  • Activates the ERK/c-Jun signaling pathway and promotes IGFBP1 protein expression in HepG2 cells.
  • Inhibits the proliferation of cancer cells SK-OV-3 and OVCAR-3.
  • Alleviates Aβ25-35-induced spatial and long-term memory impairment in mouse models.
  • Inhibits the formation of advanced glycation end-products (AGEs) and prevents the progression of diabetic nephropathy.
Catalog No. 50-004-13477 Supplier Medchemexpress LLC Supplier No. HYB1744S500UG
Pyridoxal phosphate is the active form of vitamin B6, acts as an inhibitor of reverse transcriptases, and is used for the treatment of tardive dyskinesia. This product is for research use only.
  • Severely inhibits DNA Polymerase and RNase H, leading to an immediate reduction in DNA synthesis.
  • Conjugation to pyridoxal phosphate fully inhibits NEIL2.
  • Activates the ERK/c-Jun signaling pathway and promotes IGFBP1 protein expression in HepG2 cells.
  • Inhibits the proliferation of cancer cells SK-OV-3 and OVCAR-3.
  • Alleviates Aβ25-35-induced spatial and long-term memory impairment in mouse models.
  • Inhibits the formation of advanced glycation end-products (AGEs) and prevents the progression of diabetic nephropathy.