Learn More
Selleck Chemical LLC Pyridoxal phosphate S5311-25mg
SDP
Catalog No. 503112298
Encompass_Preferred
Pyridoxal phosphate (Pyridoxal 5 -phosphate PAL-P PLP Vitamin B6) the active form of vitamin B6 acts as a coenzyme in all transamination reactions and in certain decarboxylation deamination and racemization reactions of amino acids
Catalog No. 50-311-2298 Supplier Selleck Chemical LLC Supplier No. S531125MG
Pyridoxal phosphate (Pyridoxal 5 -phosphate PAL-P PLP Vitamin B6) the active form of vitamin B6 acts as a coenzyme in all transamination reactions and in certain decarboxylation deamination and racemization reactions of amino acids