Learn More
Medchemexpress LLC Pyridoxal Phosphate-d3 | 1354560-58-9 | 98.0% | 250.16 | 500 UG
SDP
Catalog No. 5000452328
Encompass_Preferred
Pyridoxal Phosphate-d3 is the deuterium labeled Pyridoxal 5'-phosphate. Pyridoxal 5'-phosphate, the active form of vitamin B6, is an essential cofactor for multiple enzymes, including aromatic l-amino acid decarboxylase that catalyzes the final stage in the production of the neurotransmitters dopamine and serotonin. It is the most important coenzyme variant in the process of vitamin B6 intracellular phosphorylation and is interconvertible with other variants, including pyridoxine 5′‐phosphate (PNP) and pyridoxamine 5′-phosphate (PMP).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • Relevant for research in cancer, neurological, and metabolic diseases.
Catalog No. 50-004-52328 Supplier Medchemexpress LLC Supplier No. HYW011727S500UG
Pyridoxal Phosphate-d3 is the deuterium labeled Pyridoxal 5'-phosphate. Pyridoxal 5'-phosphate, the active form of vitamin B6, is an essential cofactor for multiple enzymes, including aromatic l-amino acid decarboxylase that catalyzes the final stage in the production of the neurotransmitters dopamine and serotonin. It is the most important coenzyme variant in the process of vitamin B6 intracellular phosphorylation and is interconvertible with other variants, including pyridoxine 5′‐phosphate (PNP) and pyridoxamine 5′-phosphate (PMP).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • Relevant for research in cancer, neurological, and metabolic diseases.