Learn More
AdipoGen Pyridoxal 5'-phosphate monohydrate (5 g)
SDP
Catalog No. 502054612
Encompass_Preferred
Chemical. CAS: 41468-25-1. Formula: C8H10NO6P . H2O. MW: 265.16. The enzyme cofactor Pyridoxal-5‘-phosphate monohydrate is a vitamin B6 metabolite that can modify lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase. Also serves as a coenzyme in some decarboxylation and deamination reactions. It has the ability to inhibit purinergic receptors and intracellular influx of Ca2+. Pyridoxal-5‘-phosphate monohydrate can modify peptides and suppress their precursor ionization efficiency. Research shows that pyridoxal-5‘-phosphate-dependent enzymes can be inhibited by cycloserine.
Catalog No. 50-205-4612 Supplier AdipoGen Supplier No. CDXP0461G005
Add to Cart
Add to Cart
Chemical. CAS: 41468-25-1. Formula: C8H10NO6P . H2O. MW: 265.16. The enzyme cofactor Pyridoxal-5‘-phosphate monohydrate is a vitamin B6 metabolite that can modify lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase. Also serves as a coenzyme in some decarboxylation and deamination reactions. It has the ability to inhibit purinergic receptors and intracellular influx of Ca2+. Pyridoxal-5‘-phosphate monohydrate can modify peptides and suppress their precursor ionization efficiency. Research shows that pyridoxal-5‘-phosphate-dependent enzymes can be inhibited by cycloserine.