Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridoxamine 5′-phosphate | 529-96-4 | 98.9% | 1 MG
SDP

Catalog No. 5000413465
Encompass_Preferred

Pyridoxamine 5′-phosphate is the active form of vitamin B6, bound to phosphoric acid. It acts as a co-factor for various enzymes, a central metabolite, a potent antioxidant, a vitamin B6 vitamer, and an enzyme substrate.

  • Active form of vitamin B6
  • Bound to phosphoric acid
  • Co-factor for various enzymes
  • Central metabolite
  • Potent antioxidant
  • Vitamin B6 vitamer
  • Enzyme substrate
  • Interconvertible with pyridoxal 5'-phosphate hydrate
  • Intended for research use only
  • Appearance: Solid
  • Color: White to yellow
  • Molecular weight: 248.17
  • Formula: C8H13N2O5P
  • Structure classification: Alkaloids, pyridine alkaloids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites

Catalog No. 50-004-13465 Supplier Medchemexpress LLC Supplier No. HYB17461MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridoxamine 5′-phosphate is the active form of vitamin B6, bound to phosphoric acid. It acts as a co-factor for various enzymes, a central metabolite, a potent antioxidant, a vitamin B6 vitamer, and an enzyme substrate.

  • Active form of vitamin B6
  • Bound to phosphoric acid
  • Co-factor for various enzymes
  • Central metabolite
  • Potent antioxidant
  • Vitamin B6 vitamer
  • Enzyme substrate
  • Interconvertible with pyridoxal 5'-phosphate hydrate
  • Intended for research use only
  • Appearance: Solid
  • Color: White to yellow
  • Molecular weight: 248.17
  • Formula: C8H13N2O5P
  • Structure classification: Alkaloids, pyridine alkaloids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.