Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxamine dihydrochloride, 98%, MilliporeSigma™ Supelco™

Catalog No. 111014453 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4453 5 g
11-101-4451 1 g
11-101-4452 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4453 Supplier MilliporeSigma Supelco Supplier No. P93805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridoxamine dihydrochloride is a salt of pyridoxamine, which belongs to the family of vitamin B6 compounds. Pyridoxamine occurs naturally in animal-based food products. Its deficiency in humans potentially causes sideroblastic anemia, weakness, insomnia, and neurological disorders. It has been reported to have been effective against diabetic nephropathy. Featured Industries: Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 524-36-7
Physical Form Neat
Linear Formula C8H12N2O2 · 2 HCl
MDL Number MFCD00012808
Melting Point 224°C to 226°C
Synonym 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
Type Analytical Standard
Quantity 5 g
Formula Weight 241.11
Percent Purity ≥98%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.