Learn More
Cayman Chemical Pyridoxamine (hydrochloride), Purity: ≥98%, CAS: 524-36-7, 10 g
SDP
Catalog No. 502950430
Encompass_Preferred
A 4-methylamine form of vitamin B6; scavenges isoketals in cell-free assays and inhibits the formation of lysyl-levuglandin-lactam adducts in dazoxiben-stimulated isolated human platelets at 0.1 and 1 mM; inhibits development of acellular capillaries and accumulation of CML in retina in a rat model of STZ-induced diabetes at 1 g/L in the drinking water; reverses STZ-induced deficits in the novel object recognition task in a rat model of diabetes at 0.4 and 1 g/L in the drinking water
Catalog No. 50-295-0430 Supplier Cayman Chemical Supplier No. 3389910 G
A 4-methylamine form of vitamin B6; scavenges isoketals in cell-free assays and inhibits the formation of lysyl-levuglandin-lactam adducts in dazoxiben-stimulated isolated human platelets at 0.1 and 1 mM; inhibits development of acellular capillaries and accumulation of CML in retina in a rat model of STZ-induced diabetes at 1 g/L in the drinking water; reverses STZ-induced deficits in the novel object recognition task in a rat model of diabetes at 0.4 and 1 g/L in the drinking water