Learn More
AdipoGen Pyridoxatin (250 µg)
SDP
Supplier:  AdipoGen AGCN20123C250
Encompass_Preferred
Chemical. CAS: 135529-30-5. Formula: C15H21NO3. MW: 263.3. Isolated from Albophoma yamanashiensis. Antibiotic. Free radical scavenger. Lipid peroxidation inhibitor. Antimicrobial activity against Candida albicans. Gelatinase A (MMP-2) inhibitor. Anticancer. Cytotoxic. DNA synthesis inhibitor.
Catalog No. 50-205-3492