Learn More
Abcam Pyrido x atin, MMP-2 inhibitor, 250UG
Catalog No. 502705867
Encompass_Preferred
MW 263.33 Da, Purity >95%. Free radical scavenger (IC₅₀ = 8 μM). MMP-2 inhibitor. Inhibits lipid peroxidation and DNA synthesis. Shows antifungal effects in vivo (IC₅₀ = 6 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5867 Supplier Abcam Supplier No. AB144338250UG
MW 263.33 Da, Purity >95%. Free radical scavenger (IC₅₀ = 8 μM). MMP-2 inhibitor. Inhibits lipid peroxidation and DNA synthesis. Shows antifungal effects in vivo (IC₅₀ = 6 μM).
The product is subject to the following: Abcam Restricted Use Statement