Learn More
Apexbio Technology LLC Pyridoxine 65-23-6 100mg
SDP
Supplier:  Apexbio Technology LLC B7912100
Encompass_Preferred
Pyridoxine (CAS 65-23-6) also known as vitamin B6 is a biologically active pyridine derivative widely present in food sources and studied for its cellular effects In experimental models of Alzheimer s disease pyridoxine demonstrates antioxidant activity by modulating the Nrf2/HO-1 signaling pathway thereby attenuating oxidative stress This molecule is frequently utilized in research investigating neurodegenerative disorders and oxidative damage providing insight into cellular mechanisms underlying neuronal protection
Catalog No. 50-224-0439