Learn More
Medchemexpress LLC Pyridoxine | 8059-24-3 | MFCD00006335 | 100.0% | 169.18 g/mol | C8H11NO3 | 10 MG
SDP
Catalog No. 5000286436
Encompass_Preferred
Pyridoxine (vitamin B6) is a water-soluble vitamin and enzymatic cofactor involved in amino acid, lipid, and neurotransmitter metabolism. Supplied as a high-purity research reagent, this material is intended for laboratory and analytical use and is not for human or clinical applications.
  • High-purity reagent suitable for biochemical assays and analytical methods.
  • Water-soluble vitamin and cofactor in metabolic reactions.
  • Suitable for cell studies and enzymology experiments.
  • Supplied in a 10 mg package for small-scale laboratory use.
  • Not for human consumption; intended for research use only.
Catalog No. 50-002-86436 Supplier Medchemexpress LLC Supplier No. HY15052510MG
Pyridoxine (vitamin B6) is a water-soluble vitamin and enzymatic cofactor involved in amino acid, lipid, and neurotransmitter metabolism. Supplied as a high-purity research reagent, this material is intended for laboratory and analytical use and is not for human or clinical applications.
  • High-purity reagent suitable for biochemical assays and analytical methods.
  • Water-soluble vitamin and cofactor in metabolic reactions.
  • Suitable for cell studies and enzymology experiments.
  • Supplied in a 10 mg package for small-scale laboratory use.
  • Not for human consumption; intended for research use only.