Learn More
Medchemexpress LLC Pyridoxine-d5 (Pyridoxol-d5) | 688302-31-0 | 95.0% | 1 MG
SDP
Catalog No. 5000412494
Encompass_Preferred
Pyridoxine-d5 (Pyridoxol-d5) is a deuterium labeled Pyridoxine. Pyridoxine is a pyridine derivative that exerts antioxidant effects in a cell model of Alzheimer's disease via the Nrf-2/HO-1 pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12494 Supplier Medchemexpress LLC Supplier No. HYB1328S1MG
Pyridoxine-d5 (Pyridoxol-d5) is a deuterium labeled Pyridoxine. Pyridoxine is a pyridine derivative that exerts antioxidant effects in a cell model of Alzheimer's disease via the Nrf-2/HO-1 pathway.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.