Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014757 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4757 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4757 Supplier MilliporeSigma Supelco Supplier No. PHR1036500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridoxine hydrochloride is a water soluble vitamin, widely used for the prevention of peripheral neuropathy and central nervous system effects associated with therapy involving isoniazid. It is highly required for the formation of hemoglobin and is also mainly involved in carbohydrate, fat and amino acid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

CAS 58-56-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula C8H11 NO3 · HCl
MDL Number MFCD00012807
Melting Point 214°C to 215°C
Synonym Adermine hydrochloride; PN HCl; Pyridoxol hydrochloride; Vitamin B6 hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 205.64
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.