Learn More
Medchemexpress LLC HY-B1745 100mg Medchemexpress, Pyridoxylamine CAS:85-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1745100MG
Encompass_Preferred
Medchemexpress, HY-B1745 100mg Pyridoxylamine CAS:85-87-0 Pyridoxylamine is an advanced glycation end production (AGEs) and lipoxidation end products (ALEs) inhibitor, to protect against diabetes-induced retinal vascular lesions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0161