Learn More
Selleck Chemical LLC Pyrilamine Maleate S4382-25mg
SDP
Catalog No. 503112098
Encompass_Preferred
Pyrilamine Maleate (Mepyramine)is a histamine H1 receptor inverse agonist it binds to a G protein-coupled form of the receptor and promotes a G protein-coupled inactive state of the H1 receptor that interferes with the Gq/11-mediated signaling
Catalog No. 50-311-2098 Supplier Selleck Chemical LLC Supplier No. S438225MG
Pyrilamine Maleate (Mepyramine)is a histamine H1 receptor inverse agonist it binds to a G protein-coupled form of the receptor and promotes a G protein-coupled inactive state of the H1 receptor that interferes with the Gq/11-mediated signaling