Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pyrilamine maleate

Catalog No. AAJ6324706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63247-06 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63247-06 Supplier Thermo Scientific™ Supplier No. J6324706

missing translation for 'validMainframeMsg'

Selective inverse agonist for the H-1 histamine receptor
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-33-6
Molecular Formula C{1}{7}H{2}{3}N{3}O.C{4}H{4}O{4}
MDL Number MFCD00069333
Quantity 5 g
Merck Index 14,7984
Synonym Mepyramine maleate;N-(4-Methoxyphenyl)methyl-N',N'-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt
Formula Weight 401.46
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.