Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrilamine Maleate, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605998 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-998 25 g Amber Glass Bottle
18-605-999 100 g Amber Glass Bottle
18-606-000 500 g Amber Glass Bottle
18-606-003 1 kg Amber Glass Bottle
18-606-001 5 kg Amber Glass Bottle
18-606-002 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-998 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PY10525GM
Add to Cart
Add to Cart

PY105, 59-33-6, C17H23N3O·C4H4O4

Spectrum™ Chemical Pyrilamine Maleate, USP is a first generation antihistamine, targeting the H1 receptor, used to treat the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-33-6
Molecular Formula C21H27N3O5
Molecular Weight (g/mol) 401.46
MDL Number MFCD00069333
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
IUPAC Name (2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
SMILES OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 99-103°C
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Pyrilamine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.