Learn More
Cayman Chemical PyrImethamIn 10mg
SDP
Catalog No. 502970951
Encompass_Preferred
A dihydrofolate reductase inhibitor that prevents the production of tetrahydrofolic acid in P. falciparum and other protozoa; displays a prophylactic effect (ED50 = 0.5 mg/kg) in in vivo antimalarial mouse models
Catalog No. 50-297-0951 Supplier Cayman Chemical Supplier No. 1647210 MG
A dihydrofolate reductase inhibitor that prevents the production of tetrahydrofolic acid in P. falciparum and other protozoa; displays a prophylactic effect (ED50 = 0.5 mg/kg) in in vivo antimalarial mouse models