Learn More
Cayman Chemical Pyrimethamine, Purity: ≥98%, CAS: 58-14-0, 50 mg
SDP
Catalog No. 502971932
Encompass_Preferred
A dihydrofolate reductase inhibitor that prevents the production of tetrahydrofolic acid in P. falciparum and other protozoa; displays a prophylactic effect (ED50 = 0.5 mg/kg) in in vivo antimalarial mouse models
Catalog No. 50-297-1932 Supplier Cayman Chemical Supplier No. 1647250 MG
A dihydrofolate reductase inhibitor that prevents the production of tetrahydrofolic acid in P. falciparum and other protozoa; displays a prophylactic effect (ED50 = 0.5 mg/kg) in in vivo antimalarial mouse models