Learn More
Medchemexpress LLC Pyrimethamine-d3 | 1189936-99-9 | 99.90% | 251.73 | 1 MG
SDP
Catalog No. 5000387297
Encompass_Preferred
Pyrimethamine-d3 is the deuterium labeled Pyrimethamine. Pyrimethamine is a medication used for protozoal infections; it interferes with tetrahydrofolic acid synthesis from folic acid by inhibiting the enzyme dihydrofolate reductase (DHFR).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87297 Supplier Medchemexpress LLC Supplier No. HY18062S1MG
Pyrimethamine-d3 is the deuterium labeled Pyrimethamine. Pyrimethamine is a medication used for protozoal infections; it interferes with tetrahydrofolic acid synthesis from folic acid by inhibiting the enzyme dihydrofolate reductase (DHFR).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.