Learn More
Selleck Chemical LLC Pyrimethamine 50mg 58-14-0
SDP
Catalog No. 507251
Encompass_Preferred
Pyrimethamine is a dihydrofolate reductase (DHFR) inhibitor, used as an antimalarial drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-725-1 Supplier Selleck Chemical LLC Supplier No. S200650MG
Pyrimethamine is a dihydrofolate reductase (DHFR) inhibitor, used as an antimalarial drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.