Learn More
Medchemexpress LLC HY-B2033 50mg Medchemexpress, Pyrimethanil CAS:53112-28-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB203350MG
Encompass_Preferred
Medchemexpress, HY-B2033 50mg Pyrimethanil CAS:53112-28-0 Pyrimethanil is an anilinopyrimidine and broad-spectrum contact fungicide for the control of Botrytis spp. on a wide variety of crops. Pyrimethanil inhibits the biosynthesis of methionine and other amino acids in Botrytis cinerea. Pyrimethanil can be used for the research of fungal diseases prevention on fruit, vegetable and ornamental plants with mold infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2720