Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrimethanil | 53112-28-0 | 99.9% | 200 MG
SDP

Catalog No. 5000411111
Encompass_Preferred

Pyrimethanil is an anilinopyrimidine and a broad-spectrum contact fungicide. It is used to control Botrytis species on various crops by inhibiting methionine biosynthesis in Botrytis cinerea. This compound is suitable for research aimed at preventing fungal diseases in fruit, vegetable, and ornamental plants with mold infections.

  • Inhibits methionine and other amino acids biosynthesis in Botrytis cinerea.
  • Aids research into preventing fungal diseases on fruit, vegetable, and ornamental plants.
  • Decreases polygalacturonase, cellulase, proteinase, and laccase activities.
  • Molecular weight: 199.25.
  • Molecular formula: C12H13N3.
  • Appearance: White to off-white solid.

Catalog No. 50-004-11111 Supplier Medchemexpress LLC Supplier No. HYB2033200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrimethanil is an anilinopyrimidine and a broad-spectrum contact fungicide. It is used to control Botrytis species on various crops by inhibiting methionine biosynthesis in Botrytis cinerea. This compound is suitable for research aimed at preventing fungal diseases in fruit, vegetable, and ornamental plants with mold infections.

  • Inhibits methionine and other amino acids biosynthesis in Botrytis cinerea.
  • Aids research into preventing fungal diseases on fruit, vegetable, and ornamental plants.
  • Decreases polygalacturonase, cellulase, proteinase, and laccase activities.
  • Molecular weight: 199.25.
  • Molecular formula: C12H13N3.
  • Appearance: White to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.