Learn More
Medchemexpress LLC Pyrimethanil-d5 | 2118244-83-8 | 99.0% | 1 MG
SDP
Catalog No. 5000411230
Encompass_Preferred
Pyrimethanil-d5 is the deuterium-labeled version of Pyrimethanil, an anilinopyrimidine and broad-spectrum contact fungicide. It controls Botrytis spp. on various crops by inhibiting the biosynthesis of methionine and other amino acids in *Botrytis cinerea*. It is suitable for research into fungal disease prevention in fruit, vegetable, and ornamental plants that have mold infections.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11230 Supplier Medchemexpress LLC Supplier No. HYB2033S11MG
Pyrimethanil-d5 is the deuterium-labeled version of Pyrimethanil, an anilinopyrimidine and broad-spectrum contact fungicide. It controls Botrytis spp. on various crops by inhibiting the biosynthesis of methionine and other amino acids in *Botrytis cinerea*. It is suitable for research into fungal disease prevention in fruit, vegetable, and ornamental plants that have mold infections.
  • Deuterium labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.