Learn More
FUJIFILM Biosciences Pyrimidifen Standard
Catalog No. 5000450421
Encompass_Preferred
This standard is intended for use in pesticide residue analysis It is provided as a white crystalline powder with purity greater than 98% and suitable for analytical methods including GC and NMR Store cool and away from light
Catalog No. 50-004-50421 Supplier FUJIFILM Biosciences Supplier No. 16818701
This standard is intended for use in pesticide residue analysis It is provided as a white crystalline powder with purity greater than 98% and suitable for analytical methods including GC and NMR Store cool and away from light