Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrimidin-4-yl-methanol | 33581-98-5 | >97.0% | 110.11 g/mol | C5H6N2O | 10 G
SDP

Catalog No. 5000300795
Encompass_Preferred

Pyrimidin-4-yl-methanol is a small-molecule synthetic intermediate used as a building block in medicinal and agrochemical research for preparing substituted pyrimidine derivatives.

  • Small-molecule building block for pharmaceutical synthesis.
  • Molecular formula C5H6N2O and molecular weight 110.11 g/mol.
  • Solid at room temperature, easy to handle.
  • High purity (≥97.0%) suitable for research synthesis.
  • Available in multiple laboratory pack sizes including 10 g.

Catalog No. 50-003-00795 Supplier Medchemexpress LLC Supplier No. HYW01782510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pyrimidin-4-yl-methanol is a small-molecule synthetic intermediate used as a building block in medicinal and agrochemical research for preparing substituted pyrimidine derivatives.

  • Small-molecule building block for pharmaceutical synthesis.
  • Molecular formula C5H6N2O and molecular weight 110.11 g/mol.
  • Solid at room temperature, easy to handle.
  • High purity (≥97.0%) suitable for research synthesis.
  • Available in multiple laboratory pack sizes including 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.