Learn More
Medchemexpress LLC Pyrimidine, 2-[4-(2-pyrazinyl)-1-piperazinyl]- | 137448-39-6 | 98.0% | 242.28 | 5 MG
SDP
Catalog No. 5000359319
Encompass_Preferred
HIV-1 inhibitor-47 is an inhibitor of HIV-1 that inhibits vif-dependent degradation of human APOBEC3G, with an IC50 value of 14.33 μM. This compound is also involved in the synthesis of 1-(2-pyrimidinyl)piperazine derivatives, which have potential antianxiety, antidepressant, and antipsychotic effects. It targets APOBEC3G and the HIV-1 Vif-APOBEC3G interaction.
  • Inhibits HIV-1
  • Inhibits vif-dependent degradation of human APOBEC3G
  • Potent inhibition with an IC50 value of 14.33 μM
  • Involved in 1-(2-pyrimidinyl)piperazine derivatives synthesis
  • Exhibits potential antianxiety, antidepressant, and antipsychotic effects
  • Targets APOBEC3G-apolipoprotein B mRNA editing enzyme catalytic subunit 3G (human)
  • Modulates the HIV-1 Vif-APOBEC3G interaction
Catalog No. 50-003-59319 Supplier Medchemexpress LLC Supplier No. HY1480425MG
HIV-1 inhibitor-47 is an inhibitor of HIV-1 that inhibits vif-dependent degradation of human APOBEC3G, with an IC50 value of 14.33 μM. This compound is also involved in the synthesis of 1-(2-pyrimidinyl)piperazine derivatives, which have potential antianxiety, antidepressant, and antipsychotic effects. It targets APOBEC3G and the HIV-1 Vif-APOBEC3G interaction.
  • Inhibits HIV-1
  • Inhibits vif-dependent degradation of human APOBEC3G
  • Potent inhibition with an IC50 value of 14.33 μM
  • Involved in 1-(2-pyrimidinyl)piperazine derivatives synthesis
  • Exhibits potential antianxiety, antidepressant, and antipsychotic effects
  • Targets APOBEC3G-apolipoprotein B mRNA editing enzyme catalytic subunit 3G (human)
  • Modulates the HIV-1 Vif-APOBEC3G interaction