Learn More
Medchemexpress LLC Pyrimidine-4-carboxylic acid | 31462-59-6 | 124.10 | 10 G
SDP
Catalog No. 5000445794
Encompass_Preferred
Pyrimidine-4-carboxylic acid is a biochemical reagent suitable for diverse life science research. It functions as both a biological material and an organic compound, and is also an important synthetic intermediate in pharmaceutical synthesis.
  • Biological material
  • Organic compound
  • Useful for life science related research
  • Synthetic intermediate for pharmaceutical synthesis
Catalog No. 50-004-45794 Supplier Medchemexpress LLC Supplier No. HYW00240410G
Pyrimidine-4-carboxylic acid is a biochemical reagent suitable for diverse life science research. It functions as both a biological material and an organic compound, and is also an important synthetic intermediate in pharmaceutical synthesis.
  • Biological material
  • Organic compound
  • Useful for life science related research
  • Synthetic intermediate for pharmaceutical synthesis