Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich pyrimidine-5-boronic acid

Catalog No. CDS013999 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013999 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013999 Supplier Sigma Aldrich Supplier No. CDS013999250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-aminoisoxazole (isoxazol-3-amine) is a 3-substituted isoxazole derivative. it is a structural isomer of 5-aminoisoxazole.3-aminoisoxazole (isoxazol-3-amine) may be used in the following studies:
  • As a reagent in the synthesis of n-(4-(n-isoxazol-3-ylsulfamoyl)phenyl)acetamide
  • As a starting material in the synthesis of n-(isoxazol-3-yl)-n'-(carbomethoxy)thiourea
  • As a starting material in the synthesis of (z)-2-(5-amino-1,2,4-thiadiazol-3-yl)-2-methoxy-iminoacetic acid, a side-chain of the fourth generation of cephem antibiotics

Specifications

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Linear Formula C4H5BN2O2
MDL Number MFCD03002366
Quantity 25 mL
Molecular Weight (g/mol) 123.91
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.