Learn More
Medchemexpress LLC Pyrimidine-5-boronic acid pinacol ester | 321724-19-0 | 98.0% | 206.05 | 10 G
SDP
Catalog No. 5000401414
Encompass_Preferred
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, commonly used as an intermediate in organic synthesis reactions, specifically as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and may exhibit mutagenic activity. Primarily, it is used in drug synthesis and catalytic reactions research, but it should be noted that it may also be a genotoxic impurity (GTIs).
  • Derivative of boronic acid
  • Intermediate in organic synthesis
  • Key reagent in Suzuki-Miyaura reaction
  • Generates organic free radicals
  • May exhibit mutagenic activity
  • Used in drug synthesis research
  • Used in catalytic reactions research
  • May be a genotoxic impurity
  • For research use only
Catalog No. 50-004-01414 Supplier Medchemexpress LLC Supplier No. HY4204510G
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, commonly used as an intermediate in organic synthesis reactions, specifically as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and may exhibit mutagenic activity. Primarily, it is used in drug synthesis and catalytic reactions research, but it should be noted that it may also be a genotoxic impurity (GTIs).
  • Derivative of boronic acid
  • Intermediate in organic synthesis
  • Key reagent in Suzuki-Miyaura reaction
  • Generates organic free radicals
  • May exhibit mutagenic activity
  • Used in drug synthesis research
  • Used in catalytic reactions research
  • May be a genotoxic impurity
  • For research use only