Learn More
Medchemexpress LLC Pyrimidine-5-boronic acid pinacol ester | 321724-19-0 | 98.0% | 206.05 | 25 G
SDP
Catalog No. 5000403142
Encompass_Preferred
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, an intermediate frequently used in organic synthesis reactions. It serves as a key reagent in the Suzuki-Miyaura reaction, generates organic free radicals through oxygen-mediated oxidation, and may exhibit mutagenic activity. This compound is primarily used in drug synthesis and catalytic reactions research and might be a genotoxic impurity (GTIs).
  • Key reagent in Suzuki-Miyaura reaction.
  • Used in drug synthesis.
  • Used in catalytic reactions.
  • Intended for research use only.
Catalog No. 50-004-03142 Supplier Medchemexpress LLC Supplier No. 25GHY42045
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, an intermediate frequently used in organic synthesis reactions. It serves as a key reagent in the Suzuki-Miyaura reaction, generates organic free radicals through oxygen-mediated oxidation, and may exhibit mutagenic activity. This compound is primarily used in drug synthesis and catalytic reactions research and might be a genotoxic impurity (GTIs).
  • Key reagent in Suzuki-Miyaura reaction.
  • Used in drug synthesis.
  • Used in catalytic reactions.
  • Intended for research use only.