Learn More
Medchemexpress LLC Pyrimidine-5-boronic acid pinacol ester | 321724-19-0 | 98.0% | 206.05 | 50 G
SDP
Catalog No. 5000404393
Encompass_Preferred
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid. Boronic acid is an intermediate commonly used in organic synthesis reactions and serves as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and exhibits potential mutagenic activity. Boronic acid is primarily used in research related to drug synthesis and catalytic reactions.
  • Derivative of boronic acid.
  • Intermediate commonly used in organic synthesis reactions.
  • Key reagent in the Suzuki-Miyaura reaction.
  • Generates organic free radicals through oxygen-mediated oxidation.
  • Exhibits potential mutagenic activity.
  • Primarily used in research related to drug synthesis and catalytic reactions.
  • May be a genotoxic impurity (GTIs).
Catalog No. 50-004-04393 Supplier Medchemexpress LLC Supplier No. HY4204550G
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid. Boronic acid is an intermediate commonly used in organic synthesis reactions and serves as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and exhibits potential mutagenic activity. Boronic acid is primarily used in research related to drug synthesis and catalytic reactions.
  • Derivative of boronic acid.
  • Intermediate commonly used in organic synthesis reactions.
  • Key reagent in the Suzuki-Miyaura reaction.
  • Generates organic free radicals through oxygen-mediated oxidation.
  • Exhibits potential mutagenic activity.
  • Primarily used in research related to drug synthesis and catalytic reactions.
  • May be a genotoxic impurity (GTIs).