Learn More
Medchemexpress LLC Pyrimidine-5-boronic acid pinacol ester | 321724-19-0 | 98.0% | 206.054 | 1 G
SDP
Catalog No. 5000403469
Encompass_Preferred
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, which is an intermediate commonly used in organic synthesis. It can be used as a key reagent in the Suzuki-Miyaura reaction and generates organic free radicals through oxygen-mediated oxidation. This compound exhibits potential mutagenic activity and is primarily utilized in drug synthesis and catalytic reactions research. It should be noted that it may be a genotoxic impurity (GTIs).
  • Intermediate commonly used in organic synthesis
  • Key reagent in the Suzuki-Miyaura reaction
  • Generates organic free radicals through oxygen-mediated oxidation
  • Exhibits potential mutagenic activity
  • Primarily utilized in drug synthesis and catalytic reactions research
Catalog No. 50-004-03469 Supplier Medchemexpress LLC Supplier No. HY420451G
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid, which is an intermediate commonly used in organic synthesis. It can be used as a key reagent in the Suzuki-Miyaura reaction and generates organic free radicals through oxygen-mediated oxidation. This compound exhibits potential mutagenic activity and is primarily utilized in drug synthesis and catalytic reactions research. It should be noted that it may be a genotoxic impurity (GTIs).
  • Intermediate commonly used in organic synthesis
  • Key reagent in the Suzuki-Miyaura reaction
  • Generates organic free radicals through oxygen-mediated oxidation
  • Exhibits potential mutagenic activity
  • Primarily utilized in drug synthesis and catalytic reactions research