Learn More
Medchemexpress LLC Pyrimidine-5-boronic acid pinacol ester | 321724-19-0 | 98.0% | 206.054 | 5 G
SDP
Catalog No. 5000403752
Encompass_Preferred
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid. Boronic acid is an intermediate commonly used in organic synthesis reactions and can be used as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and exhibits potential mutagenic activity. Boronic acid is mainly used in research in the fields of drug synthesis and catalytic reactions. It may also be a genotoxic impurity (GTIs).
Catalog No. 50-004-03752 Supplier Medchemexpress LLC Supplier No. HY420455G
Add to Cart
Add to Cart
Pyrimidine-5-boronic acid pinacol ester is a derivative of Boronic acid. Boronic acid is an intermediate commonly used in organic synthesis reactions and can be used as a key reagent in the Suzuki-Miyaura reaction. It generates organic free radicals through oxygen-mediated oxidation and exhibits potential mutagenic activity. Boronic acid is mainly used in research in the fields of drug synthesis and catalytic reactions. It may also be a genotoxic impurity (GTIs).