Learn More
Medchemexpress LLC Pyrimido[4,5-d]pyrimidinone derivative (1-cyclobutyl, 2,6-dimethylphenyl, 2-methoxy-4-piperazinyl) | 2172617-15-9 | 99.8% | 100 MG
SDP
Catalog No. 5000232299
Encompass_Preferred
YKL-06-061 is a potent, selective, second-generation salt-inducible kinase (SIK) inhibitor for research use. It displays low-nanomolar activity against SIK isoforms and is supplied as a purified small-molecule reagent.
  • Potent SIK inhibition (SIK1 = 6.56 nM, SIK2 = 1.77 nM, SIK3 = 20.5 nM).
  • High reported purity (approximately 99.8% by HPLC).
  • Molecular formula C30H37N7O2; molecular weight 527.66 g/mol.
  • Available in multiple sizes and formats, including milligram quantities and a 10 mM solution in DMSO.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-32299 Supplier Medchemexpress LLC Supplier No. HY120056100MG
YKL-06-061 is a potent, selective, second-generation salt-inducible kinase (SIK) inhibitor for research use. It displays low-nanomolar activity against SIK isoforms and is supplied as a purified small-molecule reagent.
  • Potent SIK inhibition (SIK1 = 6.56 nM, SIK2 = 1.77 nM, SIK3 = 20.5 nM).
  • High reported purity (approximately 99.8% by HPLC).
  • Molecular formula C30H37N7O2; molecular weight 527.66 g/mol.
  • Available in multiple sizes and formats, including milligram quantities and a 10 mM solution in DMSO.
  • For research use only; not for human or veterinary applications.