Learn More
Cayman Chemical Pyrintegrin, 1228445-38-2, 10 mg
SDP
Supplier:  Cayman Chemical 1617910
Encompass_Preferred
A 2,4-disubstituted pyrimidine that, at 2 µM, enhances the survival of hESC more than 30-fold after trypsin-mediated dissociation; increases integrin-dependent attachment of hESC to extracellular matrices
Catalog No. NC1212505
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.