Learn More
TARGETMOL CHEMICALS INC PYRINTEGRIN 5MG
SDP
Catalog No. 502581163
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pyrintegrin is an (beta)1-integrin agonist that promotes embryonic stem cells' survival. Pyrintegrin can be used as a podocyte-protective agent. Pyrintegrin enhances cell-extracellular matrix adhesion-mediated integrin signaling. purity: 97%
Catalog No. 50-258-1163 Supplier TARGETMOL CHEMICALS INC Supplier No. T166955MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pyrintegrin is an (beta)1-integrin agonist that promotes embryonic stem cells' survival. Pyrintegrin can be used as a podocyte-protective agent. Pyrintegrin enhances cell-extracellular matrix adhesion-mediated integrin signaling. purity: 97%