Learn More
STEMCELL Technologies Pyrintegrin, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 72842
Encompass
Pyrintegrin is a 2,4-disubstituted pyrimidine that  induces the activation of β1 integrin and multiple growth factor receptors, including FGFR1, IGFR1, EGFR1, and HER2 (Xu et al.).MAINTENANCE AND SELF-RENEWAL· Enhances integrin-dependent attachment and survival of human embryonic stem (ES) cells following trypsin-mediated single-cell dissociation (Xu et al.).
Catalog No. 50-197-5987
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov