Learn More
Apexbio Technology LLC Pyriproxyfen 95737-68-1 1g
SDP
Catalog No. 5000090113
Encompass_Preferred
Pyriproxyfen (CAS 95737-68-1) is a small-molecule inhibitor targeting insect juvenile hormone receptors It is designed to interfere with hormone-mediated developmental signaling thereby disrupting endocrine-regulated metamorphosis in insect larvae Pyriproxyfen exerts its biological activity primarily through binding to juvenile hormone receptors impeding physiological maturation and reproduction cycles in target arthropod populations In laboratory experiments pyriproxyfen demonstrates inhibitory activity with IC50 values ranging from approximately 0 1 to 10 M depending on insect species and experimental conditions Based on these pharmacological properties pyriproxyfen holds research potential in agricultural pest management and public health applications involving vector-borne diseases transmitted by arthropods
Catalog No. 50-000-90113 Supplier Apexbio Technology LLC Supplier No. C68671000
Pyriproxyfen (CAS 95737-68-1) is a small-molecule inhibitor targeting insect juvenile hormone receptors It is designed to interfere with hormone-mediated developmental signaling thereby disrupting endocrine-regulated metamorphosis in insect larvae Pyriproxyfen exerts its biological activity primarily through binding to juvenile hormone receptors impeding physiological maturation and reproduction cycles in target arthropod populations In laboratory experiments pyriproxyfen demonstrates inhibitory activity with IC50 values ranging from approximately 0 1 to 10 M depending on insect species and experimental conditions Based on these pharmacological properties pyriproxyfen holds research potential in agricultural pest management and public health applications involving vector-borne diseases transmitted by arthropods