Learn More
Medchemexpress LLC Pyriproxyfen | 95737-68-1 | 99.4% | 500 MG
SDP
Catalog No. 5000410848
Encompass_Preferred
Pyriproxyfen is a juvenile hormone analog and a pyridine-based pesticide. It works by preventing larvae from developing into adulthood, thus rendering them unable to reproduce. This makes it effective against a variety of arthropoda.
  • Juvenile hormone analog
  • Prevents larvae from developing into adulthood
  • Effective against various arthropoda
  • Pyridine-based pesticide
Catalog No. 50-004-10848 Supplier Medchemexpress LLC Supplier No. HYB2031500MG
Pyriproxyfen is a juvenile hormone analog and a pyridine-based pesticide. It works by preventing larvae from developing into adulthood, thus rendering them unable to reproduce. This makes it effective against a variety of arthropoda.
  • Juvenile hormone analog
  • Prevents larvae from developing into adulthood
  • Effective against various arthropoda
  • Pyridine-based pesticide