Learn More
Abcam Pyripro x yfen, larvicidal agent, 25G
Catalog No. 502706116
Encompass_Preferred
MW 321.4 Da, Purity >95%. Insect juvenile hormone mimetic. Larvicidal agent. Inhibits maturation in a broad spectrum of insect species and sterilizes adult females. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6116 Supplier Abcam Supplier No. AB14344425G
MW 321.4 Da, Purity >95%. Insect juvenile hormone mimetic. Larvicidal agent. Inhibits maturation in a broad spectrum of insect species and sterilizes adult females. Orally active.
The product is subject to the following: Abcam Restricted Use Statement