Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyripyropene A | 147444-03-9 | MFCD00929109 | 99.4% | 583.63 g/mol | C31H37NO10 | 500 UG
SDP

Catalog No. 5000228126
Encompass_Preferred

Pyripyropene A is a small-molecule research compound that potently and selectively inhibits sterol O-acyltransferase 2 (SOAT2/ACAT2). It is supplied as a 500 μg research pack intended for biochemical and cell-based studies focused on cholesterol esterification and related metabolic pathways.

  • Potent, selective SOAT2/ACAT2 inhibitor for in vitro studies.
  • High purity (≈99.4%) suitable for biochemical assays.
  • Molecular formula C31H37NO10 and molecular weight 583.63 g/mol.
  • Soluble in common organic solvents such as DMSO and methanol.
  • Includes supporting documentation: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-28126 Supplier Medchemexpress LLC Supplier No. HY117832500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyripyropene A is a small-molecule research compound that potently and selectively inhibits sterol O-acyltransferase 2 (SOAT2/ACAT2). It is supplied as a 500 μg research pack intended for biochemical and cell-based studies focused on cholesterol esterification and related metabolic pathways.

  • Potent, selective SOAT2/ACAT2 inhibitor for in vitro studies.
  • High purity (≈99.4%) suitable for biochemical assays.
  • Molecular formula C31H37NO10 and molecular weight 583.63 g/mol.
  • Soluble in common organic solvents such as DMSO and methanol.
  • Includes supporting documentation: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.