Learn More
Perkin Elmer US LLC PYRIS LN2 Dewar - 50 L
Catalog No. 502113378
Encompass_Preferred
PerkinElmer supplies a variety of accessories for DSC analyzers (with or without Autosamplers) that provide thermostatic control such as recirculating baths of coolant, sealed refrigeration units, or liquid N2 cooling systems.
Catalog No. 50-211-3378 Supplier Perkin Elmer US LLC Supplier No. N5373009
Add to Cart
Add to Cart
Request a Quote
PerkinElmer supplies a variety of accessories for DSC analyzers (with or without Autosamplers) that provide thermostatic control such as recirculating baths of coolant, sealed refrigeration units, or liquid N2 cooling systems.