Learn More
Perkin Elmer US LLC Platinum Sample Pans - Qty. 5
Catalog No. 509052228
Encompass_Preferred
For use with the TGA 8000 and Pyris 1 TGA. These platinum sample pans are also used to facilitate chemical reactions where platinum acts as a catalyst. They are easy to clean and are reusable. A crimper is not required.
Catalog No. 50-905-2228 Supplier Perkin Elmer US LLC Supplier No. N5370562
For use with the TGA 8000 and Pyris 1 TGA. These platinum sample pans are also used to facilitate chemical reactions where platinum acts as a catalyst. They are easy to clean and are reusable. A crimper is not required.