Learn More
Medchemexpress LLC Pyrithiamine hydrobromide | 534-64-5 | 96.6% | 100 MG
SDP
Catalog No. 5000232660
Encompass_Preferred
Pyrithiamine hydrobromide is the hydrobromide salt of pyrithiamine, a thiamine metabolic inhibitor used in biochemical research. It serves as a substrate for thiamine pyrophosphokinase and is applied in studies of thiamine metabolism and nervous system disorders.
  • Thiamine metabolic inhibitor used in biochemical research.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Useful in research on nervous system diseases and vitamin B1 metabolism.
  • Chemical formula C14H20Br2N4O, molecular weight 420.14 g/mol.
  • Supplied with high purity (≈96.6%).
  • Available in small research quantities, including 100 mg.
Catalog No. 50-002-32660 Supplier Medchemexpress LLC Supplier No. HY122273100MG
Pyrithiamine hydrobromide is the hydrobromide salt of pyrithiamine, a thiamine metabolic inhibitor used in biochemical research. It serves as a substrate for thiamine pyrophosphokinase and is applied in studies of thiamine metabolism and nervous system disorders.
  • Thiamine metabolic inhibitor used in biochemical research.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Useful in research on nervous system diseases and vitamin B1 metabolism.
  • Chemical formula C14H20Br2N4O, molecular weight 420.14 g/mol.
  • Supplied with high purity (≈96.6%).
  • Available in small research quantities, including 100 mg.