Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrithiamine hydrobromide | 534-64-5 | 96.6% | 25 MG
SDP

Catalog No. 5000232694
Encompass_Preferred

Pyrithiamine hydrobromide is a thiamine metabolic inhibitor supplied as a high-purity solid for laboratory research. It is used to study thiamine-dependent biochemical pathways and nervous system disease models.

  • Thiamine metabolic inhibitor for biochemical research.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Purity 96.6% (manufacturer reported).
  • Molecular formula C14H20Br2N4O and molecular weight 420.14 g·mol⁻¹.
  • CAS number 534-64-5.
  • Supplied as a 25 MG solid for laboratory use; not for human consumption.

Catalog No. 50-002-32694 Supplier Medchemexpress LLC Supplier No. HY12227325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrithiamine hydrobromide is a thiamine metabolic inhibitor supplied as a high-purity solid for laboratory research. It is used to study thiamine-dependent biochemical pathways and nervous system disease models.

  • Thiamine metabolic inhibitor for biochemical research.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Purity 96.6% (manufacturer reported).
  • Molecular formula C14H20Br2N4O and molecular weight 420.14 g·mol⁻¹.
  • CAS number 534-64-5.
  • Supplied as a 25 MG solid for laboratory use; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.