Learn More
Medchemexpress LLC Pyrithiamine hydrobromide | 534-64-5 | 96.6% | 50 MG
SDP
Catalog No. 5000231547
Encompass_Preferred
Pyrithiamine hydrobromide is a thiamine analog and metabolic inhibitor used as a research reagent to study thiamine-dependent pathways and to model thiamine deficiency in cellular and animal studies.
  • Thiamine metabolism inhibitor for experimental studies.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Suitable for biochemical and neuroscience research applications.
  • Solid, white to light yellow powder with reported purity of 96.6%.
  • Store at -20°C; follow recommended storage for solutions.
Catalog No. 50-002-31547 Supplier Medchemexpress LLC Supplier No. HY12227350MG
Pyrithiamine hydrobromide is a thiamine analog and metabolic inhibitor used as a research reagent to study thiamine-dependent pathways and to model thiamine deficiency in cellular and animal studies.
  • Thiamine metabolism inhibitor for experimental studies.
  • Acts as a substrate for thiamine pyrophosphokinase.
  • Suitable for biochemical and neuroscience research applications.
  • Solid, white to light yellow powder with reported purity of 96.6%.
  • Store at -20°C; follow recommended storage for solutions.